Tisk…

Nɑ-Acetyl-L-lysine 99+%

Dodavatel: Thermo Fisher Scientific

227630010 227632500
ACRO227630010EA 3560 CZK
ACRO227630010 ACRO227632500
Nɑ-Acetyl-L-lysine 99+%
Nalpha-Acetyl-L-lysine

Appearance: White to off-white Powder

Vzorec: C₈H₁₆N₂O₃
Bod tání: 256…258 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00008233
CAS číslo: 1946-82-3

Objednat

Výsledky specifikačního testu

Appearance White to off-white Powder
Infrared spectrum Conforms
Water ≤0.5 % (K.F.)
Specific optical rotation +5.0° to +6.0° (20°C, 589 nm) (c=1, H2O)
Titration with HClO4 ≥99.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR