Tisk…

Methacholine bromide 99%

Dodavatel: Thermo Fisher Scientific
Varování

226970050 226970250
ACRO226970050EA 2070 CZK
ACRO226970050 ACRO226970250
Methacholine bromide 99%
Methacholine bromide

Appearance: White to off-white Crystalline powder

Vzorec: C₈H₁₈BrNO₂
Bod tání: 150…152 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00011816
CAS číslo: 333-31-3

Objednat

Výsledky specifikačního testu

Appearance White to off-white Crystalline powder
Infrared spectrum Conforms
Melting point 149 to 153 °C
Titration Argentometric ≥98.5 % (on dried substance)
Loss on drying ≤1 %

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR