Tisk…

Inosine 5'-monophosphate disodium salt hydrate 97%

Dodavatel: Thermo Scientific

Synonyms: 5'-IMP 2Na hydrate, 5'-Inosinic acid disodium salt hydrate, Inosinic acid (disodium)(hydrate)(1:2:X), Inosine 5'-monophosphate disodium salt xhydrate, Disodium inosinate hydrate, Inosic acid disodium salt hydrate

226260050 226260250 226261000
ACRO226260050EA 1510 CZK
ACRO226260050 ACRO226260250 ACRO226261000
Inosine 5'-monophosphate disodium salt hydrate 97%
Inosine 5'-monophosphate disodium salt hydrate

CAS No.: 352195-40-5

Vzorec: C₁₀H₁₁N₄Na₂O₈P·xH₂O
MW: 392,17 g/mol (anhy)
Bod tání: 175 °C (rozkl.)
Storage Temperature: Refrigerator
MDL Number: MFCD00150372
CAS číslo: 352195-40-5

Objednat

Výsledky specifikačního testu

Appearance White Crystalline powder
Infrared spectrum Conforms
Water ≤28.5 % (K.F.)
UV spectrophotometry molar absorptivity 12300 +/- 4%, on dry basis
Heavy metals (as Pb) ≤20 ppm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR