Tisk…

S-(Carboxymethyl)-L-cysteine 98%

Dodavatel: Thermo Fisher Scientific
Varování

225810250 225811000 225810010 225810050
ACRO225810250EA 2900 CZK
ACRO225810250 ACRO225811000 ACRO225810010 ACRO225810050
S-(Carboxymethyl)-L-cysteine 98%
S-(Carboxymethyl)-L-cysteine

CAS No.: 638-23-3

Vzorec: C₅H₉NO₄S
Bod tání: 205…207 °C
Storage Temperature: Ambient
MDL Number: MFCD00002614
CAS číslo: 638-23-3

Objednat

Výsledky specifikačního testu

Appearance White to off-white Powder
Infrared spectrum Conforms
Loss on drying ≤0.5 % (105°C, 3 hrs)
TLC ≥97.5 %
Appearance of solution (1% soln. in 0.1 N HCl, gently warmed): clear colorless
Residue after ignition ≤0.2 %
Specific optical rotation -29° to -33° (20°C, 589 nm) (c=1, 10% NaHCO3)
Titration with HClO4 ≥97.5 %
Chloride (Cl) ≤1000 ppm
Iron (Fe) ≤5 ppm
Heavy metals (as Pb) ≤10 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR