Tisk…

D(+)-Melezitose monohydrate 99+%

Dodavatel: Thermo Fisher Scientific

Synonyms: (2R,2'R,3S,3'S,4S,4'S,5R,5'R,6R,6'R)-6,6'-(((2S,3S,4R,5R)-4-Hydroxy-2,5-bis(hydroxymethyl)tetrahydrofuran-2,3-diyl)bis(oxy))bis(2-(hydroxymethyl)tetrahydro-2H-pyran-3,4,5-triol) hydrate

225640050 225640250
ACRO225640050EA 1290 CZK
ACRO225640050 ACRO225640250
D(+)-Melezitose monohydrate 99+%
D-(+)-Melezitose monohydrate

Appearance: White Crystalline powder

Vzorec: C₁₈H₃₄O₁₇
Bod tání: 160 °C
Storage Temperature: Ambient
MDL Number: MFCD00149448
CAS číslo: 10030-67-8

Objednat

Výsledky specifikačního testu

Appearance White Crystalline powder
Infrared spectrum Conforms
Loss on drying ≤4.5 % (110°C, 1 mmHg)
Solubility (5 % in water) Clear colorless
HPLC ≥99.0 % (ELSD)
Specific optical rotation +86° to +90° (20°C, 589 nm) (c=4, water)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR