Tisk…
Petri dishes
Dodavatel: GAMA GROUP
400995 400902 400974 400900
GAMA400995EA 2600 CZK
GAMA400995 GAMA400902 GAMA400974 GAMA400900
Petri dishes
Misky kultivační Misky Petriho
Petri dishes for in vitro diagnostic use are for the cultivation of microorganisms, obtained from samples on photosensitive or solid culture media. Higher level of disinfection with peracetic acid.