Tisk…
Zinc sulphate 0.05 mol/l (0.05 N) vodný roztok USP Test Solution (TS)
Dodavatel: VWR Chemicals
85378.180EA 638 CZK
85378.180 85378.260
Zinc sulphate 0.05 mol/l (0.05 N) vodný roztok USP Test Solution (TS)
Zinc sulphate
Ready to use solutions.
  • Produced according to Pharmacopoeia requirements
  • Reduces time and expenses
  • Independent, traceable and certified
  • Conforms to USP 34
  • Certificates of analysis and safety data sheets available
Vzorec: O₄SZn
MW: 161,45 g/mol
Bod tání: 100 °C
Storage Temperature: Ambient
MDL Number: MFCD00011302
CAS číslo: 7733-02-0
EINECS: 0
ADR: 9,III
Merck Index: 12,10293