Tisk…
VWR® SMG Advanced, Váhy semi-mikro: Grafický displej
Dodavatel: VWR Collection
611-3551EA 49390 CZK
611-3551 611-3554 611-3557 611-3558 611-3561 611-3564 611-4952 611-4953 611-4954 611-4955
VWR® SMG Advanced, Váhy semi-mikro: Grafický displej
Váhy
Semi-micro balances with resolution of 0,01 mg, they are equipped with a new highly integrated electromagnetic cell. All models feature automatic internal calibration and are characterised by a high level of precision and repeatability. These VWR® semi-micro balances provide an excellent price/performance ratio, thanks to continuous intensive research and study of how weighing works. All models have a stainless steel top pan.
  • Integrated weighing sensor for enhanced stability and quick response time
  • Selectable filter levels and measuring units
  • Check weighing (alarm function), weight totalising, piece counting functions and density determination
  • Automatic internal calibration and RS232 connection as standard
  • Large graphic display available, includes GLP procedures, time/date, formulation with database capability and multiple languages
For models with internal calibration (denoted by 'i' at end of the model's name) the user can choose three ways to calibrate:
Balance will calibrate using the internal weight by pressing CAL button
Balance will automatically recalibrate when ambient temperature changes and/or after a certain time has passed (Incal+)
The user can choose to calibrate the balance with an external weight
Certifikace: EN 61326-1:2013, EN 61010-1: 2010, EN 61010-2-010:2014, EN 50581:2012
Informace o dodávce: Supplied with mains adapter and universal plug suitable for use in EU and UK and plastic in use protective cover. Optional accessories include density determination kit and serial printer.
Upozornení: Models with suffix D have dual-range, 'i' denotes internal calibration.