Tisk…
Visual colour kits, phosphate, with colour card comparator, MQuant®, Supelco®
Dodavatel: Merck
1.08046.0001 1.11138.0001 1.14449.0001 1.18465.0002 1.18466.0002 1.18388.0001
1.08046.0001EA 2610 CZK
1.08046.0001 1.11138.0001 1.14449.0001 1.18465.0002 1.18466.0002 1.18388.0001
Visual colour kits, phosphate, with colour card comparator, MQuant®, Supelco®
Kity pro environmentální testy
Quick tests, visual colour comparison.
  • Results in approximately 1 minute
  • Easy to use
  • Application-oriented measuring ranges
  • Portable
  • A range of economical refill packs (RP) are available
MQuant® with colour card comparator
By using cuvettes with a long light path, the MQuant® (MCC) test kits are especially suited for measuring low ion concentrations (some as low as 0,005 mg/l). These test kits are especially appropriate for analysing drinking water, spring water, purified and mineral water, as well as cooling and boiler water.
Informace o dodávce: Each test kit contains all the necessary reagents, reaction cuvettes, colour comparison chart and extensive instructions for use.