Tisk…

Reactive yellow 86, yellow powder

Dodavatel: MP Biomedicals

Synonyms: 1,3-Benzenedisulphonic acid, 1,3-Benzenedisulphonic acid, 4-((5-(aminocarbonyl)-1-ethyl-1,6-dihydro-2-hydroxy-4-methyl-6-oxo-3-pyridinyl)azo)-6-((4,6-dichloro-1,3,5-triazin-2-yl)amino)-, Disodium 4-[[5-(aminocarbonyl)-1-ethyl-1,6-dihydro-2-hydroxy-4-methyl-6-oxo-3-pyridyl]azo]-6-[(4,6-dichloro-1,3,5-triazin-2-yl)amino ]benzene-1,3 disulphonate

0219547925 0219547980
ICNA0219547925EA 1700 CZK
ICNA0219547925 ICNA0219547980
Reactive yellow 86, yellow powder
Reactive yellow 86
Vzorec: C₁₈H₁₄Cl₂N₈Na₂O₉S₂
MW: 667,37 g/mol
Storage Temperature: Ambient
MDL Number: MFCD02179314
CAS číslo: 61951-86-8

Objednat

Výsledky specifikačního testu

Identity Test Passes

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR