Tisk…

Phosphatidylethanol ≥98%

Dodavatel: MP Biomedicals

Synonyms: 1-Palmitoyl-2-oleoyl-sn-glycero-3-phosphoethanol, PEth-16:0/18:1

0215898310 0215898325
ICNA0215898310EA 6070 CZK
ICNA0215898310 ICNA0215898325
Phosphatidylethanol ≥98%
Phosphatidylethanol

Phosphatidylethanol is used as a chromatographic standard to assay PLD activity in intact cells via the conversion of phosphatidylcholine to phosphatidylethanol by phospholipase D in the presence of ethanol.


In the presence of ethanol, phospholipase D converts phosphatidylcholine to phosphatidylethanol. This conversion is the basis of a sensitive and selective assay for PLD activity in intact cells.


In the presence of ethanol, phospholipase D converts phosphatidylcholine to phosphatidylethanol. This conversion is the basis of a sensitive and selective assay for PLD activity in intact cells.


Soluble in chloroform.


Upozornení: Research Use Only (RUO).

Objednat

Výsledky specifikačního testu

Identity Test Passes
Purity ≥98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR