Tisk…

Fenoprofen calcium salt hydrate, white powder

Dodavatel: MP Biomedicals
Varování

Synonyms: Nalfon, Calcium-2-(m-Phenoxyphenyl)propionate hydrate, Fenopron, Nalgesic, dl-2-(3-Phenoxyphenyl)-propionic acid calcium salt hydrate, Fenoprofen calcium hydrate

0215803810 0215803805
ICNA0215803810EA 5920 CZK
ICNA0215803810 ICNA0215803805
Fenoprofen calcium salt hydrate, white powder
Fenoprofen calcium salt hydrate

Soluble in methanol (7,5 mg/ml ).


Soluble in methanol (7,5 mg/ml ).

Vzorec: C₃₀H₂₆O₆Ca·xH₂O
MW: 558,64 g/mol (anhy)
MDL Number: MFCD00242722
CAS číslo: 53746-45-5

Objednat

Výsledky specifikačního testu

Moisture content <10%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR