Tisk…

N-Acetylneuraminic acid ≥98%, powder

Dodavatel: MP Biomedicals

Synonyms: Neu5Ac, (4S,5R,6R,7S,8R)-5-Acetamido-4,6,7,8,9-pentahydroxy-2-oxononanoic acid, NANA, Sialic acid, 5-Acetamido-3,5-dideoxy-D-glycero-D-galacto-nonulopyranosonic acid, Lactaminic acid

0215385191 0215385125
ICNA0215385191EA 47030 CZK
ICNA0215385191 ICNA0215385125
N-Acetylneuraminic acid ≥98%, powder
N-Acetylneuraminic acid

Soluble in water.

Vzorec: C₁₁H₁₉NO₉
MW: 309.27 g/mol
MDL Number: MFCD00006620
CAS číslo: 131-48-6

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR