Tisk…

EDTA trisodium salt monohydrate, crystalline powder

Dodavatel: MP Biomedicals
Varování

Synonyms: Ethylenediaminetetraacetic Acid Trisodium Salt Hydrate, Trisodium hydrogen ethylenediaminetetraacetate monohydrate, Ethylenediaminetetra-acetic acid trisodium salt monohydrate, Ethylenedinitrilotetra-acetic acid trisodium salt monohydrate, Edetic acid trisodium salt monohydrate

0215108891 0215108890
ICNA0215108891EA 12780 CZK
ICNA0215108891 ICNA0215108890
EDTA trisodium salt monohydrate, crystalline powder
EDTA trisodium salt monohydrate

Ethylenediaminetetraacetic Acid Trisodium Salt Hydrate White Crystalline Powder


  • Presentation : White Crystalline Powder
  • Solubility : Water soluble. Clear colorless to light yellow solution at 32gm plus 100mL of 1M sodium hydroxide.
  • Auto Ignition : > 200 °C (DIN 51794)(Lit.)

Vzorec: C₁₀H₁₃N₂Na₃O₈·1H₂O
Storage Temperature: Ambient
CAS číslo: 10378-22-0
UN: 3077
ADR: ,III

Objednat

Výsledky specifikačního testu

Identity Test Passes


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR