Tisk…

Kyselina 4-hydroxybenzoová ≥99%, powder

Dodavatel: MP Biomedicals

0210257790 0210257791 0210257780
ICNA0210257790EA 5060 CZK
ICNA0210257790 ICNA0210257791 ICNA0210257780
Kyselina 4-hydroxybenzoová ≥99%, powder
Kyselina 4-hydroxybenzoová
Vzorec: C₇H₆O₃
MDL Number: MFCD00002547
CAS číslo: 99-96-7

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR