Tisk…

5-Iodo-2'-deoxycytidine, powder

Dodavatel: MP Biomedicals

02102062.1 02102062.5 0210206201 0210206205
ICNA02102062.1EA 1110 CZK
ICNA02102062.1 ICNA02102062.5 ICNA0210206201 ICNA0210206205
5-Iodo-2'-deoxycytidine, powder
5-Iodo-2'-deoxycytidine

5-Iodo-2′-deoxycytidine (5-iododeoxycytidine) is used in the construction of DNA oligomers to enable structural studies and photoactivated cross-linking. 5-Iodo-2′-deoxycytidine is used in the synthesis of other modified nucleosides, such as 5-ethynylferrocenyl-2′-deoxycytidine used in semiconductor electrodes and 10-(2-deoxyβ-D-ribofuranosyl)pyrimido[4′,5′:4,5]pyrimido[1,6-a]indole-6,9(7H)-dione (dCPPI).

Vzorec: C₉H₁₂IN₃O₄
MDL Number: MFCD00038063
CAS číslo: 611-53-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR