Tisk…

2-Keto-D-gluconic acid hemicalcium salt monohydrate ≥98%, powder

Dodavatel: MP Biomedicals
Varování

0210036505 0210036501
ICNA0210036505EA 16860 CZK
ICNA0210036505 ICNA0210036501
2-Keto-D-gluconic acid hemicalcium salt monohydrate ≥98%, powder
2-Keto-D-gluconic acid hemicalcium salt monohydrate

2-Keto-D-gluconic Acid hemicalcium salt is Hemicalcium Salt of 2-Keto-D-gluconic acid. It is produced from D-glucose.
Presentation: White Powder


2-Keto-D-gluconic Acid hemicalcium salt is Hemicalcium Salt of 2-Keto-D-gluconic acid. It is produced from D-glucose.

Vzorec: C₆H₉O₇·½Ca·H₂O
Storage Temperature: Refrigerator
MDL Number: MFCD00149104
CAS číslo: 3470-37-9

Objednat

Výsledky specifikačního testu

Identity Test Passes
Purity 99.3%
FTIR Conforms to Standard
Optical Rotation -67.2°
Solubility (5% aq soln) Clear, Colorless Solution


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR