N-Acetyl-L-arginine dihydrate, powder
Dodavatel: MP Biomedicals
0210007805
0210007810
ICNA0210007805EA
2180
CZK
ICNA0210007805
ICNA0210007810
N-Acetyl-L-arginine dihydrate, powder
N-Acetyl-L-arginine dihydrate
Clear colorless solution at 50 mg/mL in 1 M hydrochloric acid.
Vzorec:
C₈H₁₆N₄O₃·₂H₂O |
MDL Number:
MFCD00019733 CAS číslo: 210545-23-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...