Cytosine-β-D-arabinofuranoside hydrochloride, powder
Dodavatel: MP Biomedicals
|
Varování
|
Synonyms:
Arabinosylcytosine hydrochloride, Cytarabine HCI, Cytarabine HCl, 4-Amino-1-((2S,3R,4R,5S)-3,4-dihydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)pyrimidin-2(1H)-one hydrochloride, 1-(β-D-Arabinofuranosyl)cytosine HCl
Soluble in water (50 mg/ml - clear, colourless solution).
Cytosine β-D-arabinofuranoside hydrochloride is an anti-leukemic agent. It results in DNA fragmentation and inhibition of replication. In addition this compound does not have any effects on RNA synthesis. Cytosine β-D-arabinofuranoside hydrochloride can function as a ligand which shows strong affinity and selectivity for the Translocator protein (TSPO). Studies suggest that cytosine β-D-arabinofuranoside hydrochloride can down-regulate deoxycytidine kinase in acute myeloid leukemia murine cell lines which inhibits the phosphorylation of several deoxyribonucleosides. Furthermore, cytosine β-D-arabinofuranoside hydrochloride can be inhibited in myeloid leukemic cells via suppression of ENT1 promoter activity, responsible for transporting cytosine β-D-arabinofuranoside into the cell.
Cytosine β-D-arabinofuranoside hydrochloride is a selective inhibitor of DNA synthesis that does not affect RNA synthesis in mammalian cells. It is used as an antineoplastic and antiviral agent.
Ara-C an anti-leukemia agent is phosphorylated to Ara-CTP and is incorporatee into DNA. It inhibits DNA replication by forming cleavage complexes with topoisomerase I resulting in DNA fragmentation, and ultimately induces apoptosis via the PKC signaling pathway. It does not inhibit RNA synthesis.
Vzorec:
C₉H₁₄ClN₃O₅ MW: 279,677 g/mol |
MDL Number:
MFCD00012839 CAS číslo: 69-74-9 |
Výsledky specifikačního testu
Form | White Powder |
Storage | Store at 2 to 8 °C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...