Tisk…

Uhličitan vápenatý, light powder USP

Dodavatel: Spectrum Chemical

CA130-12KG CA130-2.5KG CA130-45KGBL CA130-500GM
SPCMCA130-12KGEA 20900 CZK
SPCMCA130-12KG SPCMCA130-2.5KG SPCMCA130-45KGBL SPCMCA130-500GM
Uhličitan vápenatý, light powder USP
Uhličitan vápenatý

CALCIUM CARBONATE, LIGHT POWDER, USP is a naturally found common substance found in rocks and it is the main component of shells of marine organisms, snails, coal balls, pearls, and eggshells. Calcium carbonate is used as a calcium supplement and as an antacid however excessive consumption can be hazardous. The USP grade indicates it is graded suitable for personal care, cosmetic and pharmaceutical applications.


All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
BSE/TSE statement (available upon request)
Allergen statement (available upon request)
Kosher certified
Storage temperature - HYGROSCOPIC: Keep tightly closed.

Vzorec: CaCO₃
Bod tání: 825 °C
Hustota: 2,93 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010906
CAS číslo: 471-34-1
EINECS: 207-439-9
Merck Index: 13,01658

Objednat

Výsledky specifikačního testu

Assay 98% Min.

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR