Tisk…
Visual colour kits, copper, MQuant®, Supelco®
Dodavatel: Merck
1.18459.0002 1.14765.0001
1.18459.0002EA 5990 CZK
1.18459.0002 1.14765.0001
Visual colour kits, copper, MQuant®, Supelco®
Kity pro environmentální testy
Quick tests for visual colour comparison.
  • Results in approximately five minutes
  • Easy to use
  • Application-oriented measuring ranges
  • Portable
  • A range of economical refill packs (RP) are available
MQuant® with colour disc comparator
Using the comparator disc, these test kits are especially suitable for the analysis of turbid and coloured waters. The comparator disc is a practical and robust colour comparison device that provides reliable measurements, even in humid environments. Ideal for samples of medium to high concentration in applications ranging from testing bottled water to wastewater and other industrial water testing.
RP= Refill pack.
Informace o dodávce: Each test kit contains all the necessary reagents, reaction cuvettes, colour comparison chart and extensive instructions for use.