Tisk…

Xantanová guma NF

Dodavatel: Spectrum Chemical

Synonyms: Xanthan

XA105-100GM XA105-12KG XA105-2.5KG XA105-500GM XA105-45KGBL
SPCMXA105-100GMEA 2790 CZK
SPCMXA105-100GM SPCMXA105-12KG SPCMXA105-2.5KG SPCMXA105-500GM SPCMXA105-45KGBL
Xantanová guma NF
Xantanová guma

Xanthan Gum, NF is a natural gum polysaccharide used as a thickener or rheology modifier and emulsion stabilizer. Spectrum Chemical NF products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities

Hustota: 1,5 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00131256
CAS číslo: 11138-66-2
EINECS: 234-394-2

Objednat

Výsledky specifikačního testu

Assay 91% Min.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR