Brij® S100
Dodavatel: Spectrum Chemical
Synonyms:
Polyoxyethylene (100) stearyl ether
B1593-2.5KG
B1593-25GM
SPCMB1593-2.5KGEA
43200
CZK
SPCMB1593-2.5KG
SPCMB1593-25GM
Brij® S100
Brij® S100
Vzorec:
C₁₈H₃₇(OCH₂CH₂)nOH,n~100 |
MDL Number:
MFCD00074177 CAS číslo: 9005-00-9 EINECS: 500-017-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...