Starch (from corn), powder NF
Dodavatel: Spectrum Chemical
S1552-12KG
S1552-45KGBL
S1552-500GM
SPCMS1552-12KGEA
21080
CZK
SPCMS1552-12KG
SPCMS1552-45KGBL
SPCMS1552-500GM
Starch (from corn), powder NF
Starch
Melamine statement (available upon request)
BSE/TSE statement (available upon request)
Vzorec:
(C₆H₁₀O₅)n Storage Temperature: Ambient |
MDL Number:
MFCD00082026 CAS číslo: 9005-25-8 EINECS: 232-679-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...