Tisk…
Polyvinylpyrrolidone, M.W. ∼ 40,000 USP (K30)
Dodavatel: Spectrum Chemical
Synonyms: Povidone, Polyvinylpyrrolidon, Polyvidone, PVP
P1454-50KGBL P1454-5KG P1454-12KGBL
SPCMP1454-50KGBLEA 0 CZK
SPCMP1454-50KGBL SPCMP1454-5KG SPCMP1454-12KGBL
Polyvinylpyrrolidone, M.W. ∼ 40,000 USP (K30)
Polyvinylpyrrolidone
Melamine statement (available upon request)
BSE/TSE statement (available upon request)
Vzorec: (C₆H₉NO)n
Storage Temperature: Ambient
MDL Number: MFCD01076626
CAS číslo: 9003-39-8