DL-Menthol, crystals USP
Dodavatel: Spectrum Chemical
Synonyms:
rel-(1R,2S,5R)-2-Isopropyl-5-methylcyclohexanol
ME111-10KGBL
ME111-180KGBLK
ME111-50KGBL
ME111-500GM
SPCMME111-10KGBLEA
114280
CZK
SPCMME111-10KGBL
SPCMME111-180KGBLK
SPCMME111-50KGBL
SPCMME111-500GM
DL-Menthol, crystals USP
DL-Menthol
Vzorec:
C₁₀H₂₀O MW: 156,27 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00001484 CAS číslo: 89-78-1 EINECS: 201-939-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...