Tisk…

Dipropyleneglycol (mixture of isomers)

Dodavatel: Spectrum Chemical

Synonyms: Dipropylene glycol

D1207-200LTBL D1207-20LT
SPCMD1207-200LTBLEA 100800 CZK
SPCMD1207-200LTBL SPCMD1207-20LT
Dipropyleneglycol (mixture of isomers)
Dipropyleneglycol (mixture of isomers)
Vzorec: C₆H₁₄O₃
MW: 134,18 g/mol
MDL Number: MFCD00051023
CAS číslo: 25265-71-8
EINECS: 246-770-3

Objednat

Výsledky specifikačního testu

Assay 99% Min.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR