4-Nitrophenylhydrazine hydrochloride ≥98%
Dodavatel: Thermo Scientific
|
Varování
|
Synonyms:
1-(4-Nitrophenyl)hydrazine hydrochloride, (4-Nitrophenyl)hydrazine hydrochloride, 4-Nitrophenylhydrazinium chloride
L09273.09
L09273.04
L09273.09EA
4290
CZK
L09273.09
L09273.04
4-Nitrophenylhydrazine hydrochloride ≥98%
4-Nitrophenylhydrazine hydrochloride
4-Nitrophenylhydrazine mono and dihydrochloride exhibits potential anticancer activity. It acts as a derivatizing agent for carbonyl compounds. It is also used as pharmaceutical intermediates.
Purity: 98%
Molecular Formula: C6H7N3O2.xHCl
Formula Weight: 153.14(f.b.)
Melting Point: ca 200° dec.
UN No.: 2811
MDL No.: MFCD00040592
Vzorec:
C₆H₇N₃O₂·HCl MW: 189,6 g/mol Bod tání: ∼ 200 °C (rozkl.) Storage Temperature: Ambient |
MDL Number:
MFCD00040592 CAS číslo: 636-99-7 EINECS: 211-273-2 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...