Tisk…

L-Prolinamide 98%

Dodavatel: Thermo Fisher Scientific

Synonyms: H-Pro-NH₂, (S)-2-Pyrrolidinecarboxamide, H-Pro-NH2

L20417.03 L20417.06
L20417.03EA 2180 CZK
L20417.03 L20417.06
L-Prolinamide 98%
L-Prolinamide

Purity: 99% CAS No.: 7531-52-4 Molecular Formula: C5H10N2O Formula Weight: 114.15 Melting Point: 95-99° Beilstein Reference: 80807 Optical Rotation: -105° (c=2 in ethanol) MDL No.: MFCD00005253

Vzorec: C₅H₁₀N₂O
MW: 114.15 g/mol
Bod tání: 95…99 °C
Storage Temperature: Ambient
MDL Number: MFCD00005253
CAS číslo: 7531-52-4
EINECS: 231-397-0

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR