2-(Trifluoromethyl)phenylglyoxal hydrate 98% (dry weight)
Dodavatel: Thermo Scientific
|
Varování
|
L20280.03
L20280.MD
L20280.03EA
4530
CZK
L20280.03
L20280.MD
2-(Trifluoromethyl)phenylglyoxal hydrate 98% (dry weight)
2-(Trifluoromethyl)phenylglyoxal hydrate
Purity: 98%
CAS No.: 745783-91-9
Molecular Formula: C9H5F3O2.xH2O
Formula Weight: 202.13(anhy)
Melting Point: 36-43°
MDL No.: MFCD05664102
Vzorec:
C₉H₇F₃O₃ Bod tání: 36…43 °C Storage Temperature: Ambient |
MDL Number:
MFCD06801323 CAS číslo: 745783-91-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...