4-Methoxyphenylglyoxal hydrate 95% (dry weight)
Dodavatel: Thermo Scientific
|
Varování
|
L20274.03
L20274.06
L20274.03EA
2350
CZK
L20274.03
L20274.06
4-Methoxyphenylglyoxal hydrate 95% (dry weight)
4-Methoxyphenylglyoxal hydrate
Purity: 95%
CAS No.: 16208-17-6
Molecular Formula: C9H8O3.xH2O
Formula Weight: 164.16(anhy)
Melting Point: 104-111°
Boiling Point: 145-147°/13mm
Beilstein Reference: 606701
MDL No.: MFCD00234534
Vzorec:
C₉H₁₀O₄ Bod varu: 145…147 °C (13 mmHg) Bod tání: 104…111 °C Storage Temperature: Ambient |
MDL Number:
MFCD00234534 CAS číslo: 16208-17-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...