Tisk…

Ethyl-(S)-(+)-4-cyano-3-hydroxybutyrate ≥97%, ee 98%

Dodavatel: Thermo Scientific
Varování

Synonyms: Ethyl (S)-4-cyano-3-hydroxybutyrate

L20259.06 L20259.14
L20259.06EA 3470 CZK
L20259.06 L20259.14
Ethyl-(S)-(+)-4-cyano-3-hydroxybutyrate ≥97%, ee 98%
Ethyl-(S)-(+)-4-cyano-3-hydroxybutyrate

Purity: 97% CAS No.: 312745-91-8 Molecular Formula: C7H11NO3 Formula Weight: 157.17 Flash Point: 110°(230°F) Refractive Index: 1.4480 Boiling Point: 270° Optical Rotation: +5°(neat) MDL No.: MFCD00672878

Vzorec: C₇H₁₁NO₃
MW: 157,17 g/mol
Bod varu: 270 °C
Hustota: 1,114
Bod vzplanutí: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00672878
CAS číslo: 312745-91-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR