Tisk…

2-Nitrophenyl selenocyanate 97%

Dodavatel: Thermo Fisher Scientific
Nebezpečí

L20193.03 L20193.06
L20193.03EA 1780 CZK
L20193.03 L20193.06
2-Nitrophenyl selenocyanate 97%
2-Nitrophenyl selenocyanate

Purity: 97% CAS No.: 51694-22-5 Molecular Formula: C7H4N2O2Se Formula Weight: 227.08 Melting Point: 134-141° Beilstein Reference: 1309777 UN No.: UN3283 MDL No.: MFCD00043146

Vzorec: C₇H₄N₂O₂Se
Bod tání: 134…141 °C
Storage Temperature: Ambient
MDL Number: MFCD00043146
CAS číslo: 51694-22-5
EINECS: 257-350-4
ADR: 6.1,II

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR