2-(Aminomethyl)piperidine 98%
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
L20122.06
L20122.14
L20122.06EA
1480
CZK
L20122.06
L20122.14
2-(Aminomethyl)piperidine 98%
2-(Aminomethyl)piperidine
Purity: 98%
CAS No.: 22990-77-8
Molecular Formula: C6H14N2
Formula Weight: 114.19
Flash Point: 68°(154°F)
Refractive Index: 1.4850
Boiling Point: 63-66°/12mm
Beilstein Reference: 102994
UN No.: UN2735
MDL No.: MFCD00129011
Vzorec:
C₆H₁₄N₂ Bod varu: 63…66 °C (12 mmHg) Hustota: 0,942 Bod vzplanutí: 68 °C (154 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00129011 CAS číslo: 22990-77-8 EINECS: 245-371-1 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...