Tisk…

Fmoc-1-aminocyclohexane-1-carboxylic acid ≥98%

Dodavatel: Thermo Fisher Scientific

L19810.03 L19810.06
L19810.03EA 2250 CZK
L19810.03 L19810.06
Fmoc-1-aminocyclohexane-1-carboxylic acid ≥98%
Fmoc-1-aminocyclohexane-1-carboxylic acid

Purity: 98+% CAS No.: 162648-54-6 Molecular Formula: C22H23NO4 Formula Weight: 365.43 Storage Temperature: 2°C to 8°C Melting Point: 185-187° dec. Beilstein Reference: 7887646 MDL No.: MFCD00273464

Vzorec: C₂₂H₂₃NO₄
MDL Number: MFCD00273464
CAS číslo: 162648-54-6

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR