(3R)-(-)-1-Benzyl-3-aminopyrrolidine 99%, ee 99%
Dodavatel: Thermo Scientific
|
Nebezpečí
|
Synonyms:
(R)-1-Benzylpyrrolidin-3-amine, (R)-(-)-1-Benzyl-3-aminopyrrolidine
L19690.03
L19690.06
L19690.03EA
1420
CZK
L19690.03
L19690.06
(3R)-(-)-1-Benzyl-3-aminopyrrolidine 99%, ee 99%
(3R)-(-)-1-Benzyl-3-aminopyrrolidine
Purity: 99%
CAS No.: 114715-39-8
Molecular Formula: C11H16N2
Formula Weight: 176.26
Flash Point: 140°(284°F)
Refractive Index: 1.5430
Boiling Point: 101-103°/3mm
Beilstein Reference: 6477592
Optical Rotation: -2° (neat)
UN No.: UN2735
MDL No.: MFCD00082638
Vzorec:
C₁₁H₁₆N₂ MW: 176,26 g/mol Bod varu: 101…103 °C (3 mmHg) Hustota: 1,020 Bod vzplanutí: 140 °C (284 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00082638 CAS číslo: 114715-39-8 UN: 2735 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...