Tisk…

Amberlite® IRC-748 ion exchange resin

Dodavatel: Thermo Fisher Scientific

Synonyms: Amberlite® IRC-748 IRC-748, Amberlite® IRC-748

L19570.30 L19570.0B
L19570.30EA 1550 CZK
L19570.30 L19570.0B
Amberlite® IRC-748 ion exchange resin
Amberlite® IRC-748

CAS No.: 79620-28-3 Odour: Odourless MDL No.: MFCD00132702

Storage Temperature: Ambient
MDL Number: MFCD00132702
CAS číslo: 79620-28-3
Merck Index: 14,382

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR