Tisk…

Amberlite® IRA-900 (Cl) ion exchange resin

Dodavatel: Thermo Fisher Scientific

Synonyms: Amberlite®, IRA-900 chloride form, Amberlite® IRA-900

L19567.30 L19567.0B
L19567.30EA 1600 CZK
L19567.30 L19567.0B
Amberlite® IRA-900 (Cl) ion exchange resin
Amberlite® IRA-900 (Cl)

CAS No.: 9050-97-9 Odour: Odourless MDL No.: MFCD00132712

Storage Temperature: Ambient
MDL Number: MFCD00132712
CAS číslo: 9050-97-9
Merck Index: 14,382

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR