Tisk…

4-(Methylsulphonyl)phenylacetic acid 97%

Dodavatel: Thermo Fisher Scientific
Varování

L19504.06 L19504.14
L19504.06EA 1480 CZK
L19504.06 L19504.14
4-(Methylsulphonyl)phenylacetic acid 97%
4-(Methylsulphonyl)phenylacetic acid

Purity: 97% CAS No.: 90536-66-6 Molecular Formula: C9H10O4S Formula Weight: 214.24 Melting Point: 134-138° Beilstein Reference: 2646649 MDL No.: MFCD00216495

Vzorec: C₉H₁₀O₄S
Bod tání: 134…138 °C
Storage Temperature: Ambient
MDL Number: MFCD00216495
CAS číslo: 90536-66-6

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR