Tisk…

5-Bromo-2-methyl-2-pentene 97%

Dodavatel: Thermo Scientific
Varování

Synonyms: 5-Bromo-2-methylpent-2-ene

L19406.03 L19406.06
L19406.03EA 2360 CZK
L19406.03 L19406.06
5-Bromo-2-methyl-2-pentene 97%
5-Bromo-2-methyl-2-pentene

Purity: 97% CAS No.: 2270-59-9 Molecular Formula: C6H11Br Formula Weight: 163.06 Storage Temperature: 2°C to 8°C Flash Point: 23°(73°F) Refractive Index: 1.4775 Boiling Point: 50°/20mm Beilstein Reference: 1735749 UN No.: UN1993 MDL No.: MFCD00009887

Vzorec: C₆H₁₁Br
MW: 163,06 g/mol
Bod varu: 50 °C (20 mmHg)
Hustota: 1,219
Bod vzplanutí: 23 °C (73 °F)
MDL Number: MFCD00009887
CAS číslo: 2270-59-9
ADR: 3,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR