(R)-2-Amino-1-phenylethanol 97%, ee 98%
Dodavatel: Thermo Scientific
|
Nebezpečí
|
Synonyms:
(R)-(-)-2-Amino-1-phenylethanol, (R)-(-)-alpha-(Aminomethyl)benzyl alcohol
L19403.06
L19403.03
L19403.06EA
7900
CZK
L19403.06
L19403.03
(R)-2-Amino-1-phenylethanol 97%, ee 98%
(R)-2-Amino-1-phenylethanol
Purity: 97%
CAS No.: 2549-14-6
Molecular Formula: C8H11NO
Formula Weight: 137.18
Odour: Amine-like
Melting Point: 57-63°
Boiling Point: 160°/17mm
Beilstein Reference: 3196196
Optical Rotation: -43° (c=2 in ethanol)
UN No.: UN3259
MDL No.: MFCD00239406
Vzorec:
C₈H₁₁NO MW: 137,18 g/mol Bod varu: 160 °C (17 mmHg) Bod tání: 57…63 °C Storage Temperature: Ambient |
MDL Number:
MFCD00239406 CAS číslo: 2549-14-6 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...