Tisk…

(S)-1-(tert-Butoxycarbonyl)piperidine-2-carboxylic acid ≥98%

Dodavatel: Thermo Fisher Scientific
Varování

Synonyms: Boc-Homopro-OH, 1-(tert-Butoxycarbonyl)-L-pipecolic Acid, Boc-HomoPro-OH, 1-Boc-L-pipecolic Acid, (S)-1-Boc-2-piperidinecarboxylic Acid, Boc-Pip-OH, (S)-N-BOC-Piperidine-2-carboxylic acid

L19361.03 L19361.06
L19361.03EA 1070 CZK
L19361.03 L19361.06
(S)-1-(tert-Butoxycarbonyl)piperidine-2-carboxylic acid ≥98%
(S)-1-(tert-Butoxycarbonyl)piperidine-2-carboxylic acid

Purity: 98+% CAS No.: 26250-84-0 Molecular Formula: C11H19NO4 Formula Weight: 229.28 Melting Point: 121-125° Beilstein Reference: 3652038 Optical Rotation: -46° (c=1 in methanol) MDL No.: MFCD00151904

Vzorec: C₁₁H₁₉NO₄
Bod tání: 121…125 °C
Storage Temperature: Ambient
MDL Number: MFCD00151904
CAS číslo: 26250-84-0

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR