Tisk…

1-Benzylpiperidine 98%

Dodavatel: Thermo Scientific
Varování

L19339.09 L19339.18
L19339.09EA 2980 CZK
L19339.09 L19339.18
1-Benzylpiperidine 98%
1-Benzylpiperidine

Purity: 98% CAS No.: 2905-56-8 Molecular Formula: C12H17N Formula Weight: 175.28 Refractive Index: 1.5260 Boiling Point: 120-123°/9mm Beilstein Reference: 5639 MDL No.: MFCD00224901

Vzorec: C₁₂H₁₇N
MW: 175,28 g/mol
Bod varu: 120…123 °C (9 mmHg)
Hustota: 0,950
Storage Temperature: Ambient
MDL Number: MFCD00224901
CAS číslo: 2905-56-8
EINECS: 220-809-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR