5-Methoxy-1H-indole-3-carbaldehyde 99%
Dodavatel: Thermo Scientific
|
Varování
|
L19072.03
L19072.06
L19072.03EA
2840
CZK
L19072.03
L19072.06
5-Methoxy-1H-indole-3-carbaldehyde 99%
5-Methoxy-1H-indole-3-carbaldehyde
Purity: 99%
CAS No.: 10601-19-1
Molecular Formula: C10H9NO2
Formula Weight: 175.19
Melting Point: 179-183°
Beilstein Reference: 132769
MDL No.: MFCD00005623
Vzorec:
C₁₀H₉NO₂ MW: 175,19 g/mol Bod tání: 179…183 °C Storage Temperature: Ambient |
MDL Number:
MFCD00005623 CAS číslo: 10601-19-1 EINECS: 234-220-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...