Tisk…

Ambersep® 900 (OH) ion exchange resin

Dodavatel: Thermo Fisher Scientific

L19035.22 L19035.36 L19035.0B
L19035.22EA 1080 CZK
L19035.22 L19035.36 L19035.0B
Ambersep® 900 (OH) ion exchange resin
Ambersep® 900 (OH)

CAS No.: 9017-79-2 Odour: Odourless MDL No.: MFCD00802695

Storage Temperature: Ambient
MDL Number: MFCD00802695
CAS číslo: 9017-79-2

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR