Tisk…

4-Isopropoxybenzenesulphonyl chloride 96%

Dodavatel: Thermo Fisher Scientific
Nebezpečí

L18984.03 L18984.06
L18984.03EA 2390 CZK
L18984.03 L18984.06
4-Isopropoxybenzenesulphonyl chloride 96%
4-Isopropoxybenzenesulphonyl chloride

Purity: 96% CAS No.: 98995-40-5 Molecular Formula: C9H11ClO3S Formula Weight: 234.70 Physical Form: Crystalline solid Sensitivity: Moisture Sensitive Appearance: White to pale cream Melting Point: 30-37° Boiling Point: 112-113°/0.01mm Beilstein Reference: 3269058 UN No.: UN3261 MDL No.: MFCD03093083

Vzorec: C₉H₁₁ClO₃S
Bod varu: 112…113 °C (0,01 mmHg)
Bod tání: 30…37 °C
Storage Temperature: Ambient
MDL Number: MFCD03093083
CAS číslo: 98995-40-5
ADR: 8,II

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR