1,1,3,3-Tetramethylbutyl isocyanate 98%
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
Purity: 98%
CAS No.: 1611-57-0
Molecular Formula: C9H17NO
Formula Weight: 155.24
Storage Temperature: 2°C to 8°C
Flash Point: 51°(123°F)
Refractive Index: 1.4260
Odour: Pungent
Boiling Point: 170-172°
UN No.: UN3080
MDL No.: MFCD00209615
Vzorec:
C₉H₁₇NO Bod varu: 170…172 °C Hustota: 0,859 Bod vzplanutí: 51 °C (123 °F) |
MDL Number:
MFCD00209615 CAS číslo: 1611-57-0 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...