Tisk…

Ethyl-(S)-4-Chloro-3-hydroxybutyrate 98%

Dodavatel: Thermo Scientific
Nebezpečí

Synonyms: (S)-4-Chloro-3-hydroxybutyric Acid Ethyl Ester, Ethyl (S)-4-chloro-3-hydroxybutanoate

L18554.03 L18554.06
L18554.03EA 501 CZK
L18554.03 L18554.06
Ethyl-(S)-4-Chloro-3-hydroxybutyrate 98%
Ethyl-(S)-4-Chloro-3-hydroxybutyrate

Purity: 98% CAS No.: 86728-85-0 Molecular Formula: C6H11ClO3 Formula Weight: 166.61 Storage Temperature: 2°C to 8°C Flash Point: 110°(230°F) Refractive Index: 1.4530 Boiling Point: 93-95°/5mm Optical Rotation: -14° (neat) MDL No.: MFCD00211241

Vzorec: C₆H₁₁ClO₃
MW: 166,60 g/mol
Bod varu: 93…95 °C (5 mmHg)
Hustota: 1,190
Bod vzplanutí: >110 °C (230 °F)
MDL Number: MFCD00211241
CAS číslo: 86728-85-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR