N-(tert-Butoxycarbonyl)-D-2-phenylglycine 99%
Dodavatel: Thermo Scientific
Synonyms:
N-Boc-D-2-phenylglycine, BOC-D-alpha-phenylglycine, Boc-D-Phg-OH
L18540.03
L18540.06
L18540.03EA
1260
CZK
L18540.03
L18540.06
N-(tert-Butoxycarbonyl)-D-2-phenylglycine 99%
N-(tert-Butoxycarbonyl)-D-2-phenylglycine
Purity: 99%
CAS No.: 33125-05-2
Molecular Formula: C13H17NO4
Formula Weight: 251.28
Storage Temperature: 2°C to 8°C
Melting Point: 90-92°
Beilstein Reference: 3033982
Optical Rotation: -144° (c=1 in ethanol)
MDL No.: MFCD00062043
Vzorec:
C₁₃H₁₇NO₄ MW: 251,28 g/mol Bod tání: 90…92 °C |
MDL Number:
MFCD00062043 CAS číslo: 33125-05-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...