(S)-(+)-5-Hydroxymethyl-2-pyrrolidinone 98%
Dodavatel: Thermo Scientific
|
Varování
|
L18359.06
L18359.03
L18359.06EA
4230
CZK
L18359.06
L18359.03
(S)-(+)-5-Hydroxymethyl-2-pyrrolidinone 98%
(S)-(+)-5-Hydroxymethyl-2-pyrrolidinone
Purity: 98%
CAS No.: 17342-08-4
Molecular Formula: C5H9NO2
Formula Weight: 115.13
Melting Point: 78-82°
Boiling Point: 147-149°/0.06m
Beilstein Reference: 4657914
Optical Rotation: +30° (c=2 in ethanol)
MDL No.: MFCD00077792
Vzorec:
C₅H₉NO₂ MW: 115,13 g/mol Bod varu: 147…149 °C (0,06m) Bod tání: 78…82 °C Storage Temperature: Ambient |
MDL Number:
MFCD00077792 CAS číslo: 17342-08-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...